Tin(IV) fluoride ≥98.5%

Supplier: STREM CHEMICALS INC MS
Danger

93-5024-5G 93-5024-1G
100198-406EA 225.05 USD
100198-406 100196-706
Tin(IV) fluoride ≥98.5%
Tin (IV) fluoride
Formula: SnF₄
Boiling Pt: 705 °C (1013 hPa)
Melting Pt: 705 °C (subl.)
Density: 4.780
Storage Temperature: Ambient
MDL Number: MFCD00042541
CAS Number: 7783-62-2
EINECS: 232-016-0
UN: 1759
ADR: 8,II
Merck Index: 12,08931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR