Silver 2-ethylhexanoate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-4741-1G 93-4741-5G
100196-686EA 67.94 USD
100196-686 100210-420
Silver 2-ethylhexanoate ≥99%
Silver 2-ethylhexanoate
Formula: C₈H₁₅AgO₂
MDL Number: MFCD00058700
CAS Number: 26077-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR