Niobium(V) ethanolate ≥99.9% (Nb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Niobium(V) ethoxide
93-4104-5G
93-4104-25G
100196-656EA
101.91
USD
100196-656
100203-468
Niobium(V) ethanolate ≥99.9% (Nb basis)
Niobium(V) ethanolate
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formula:
C₁₀H₂₅NbO₅ Boiling Pt: 142 °C (0.1 mmHg) Melting Pt: 6 °C Density: 1,32 |
MDL Number:
MFCD00015122 CAS Number: 3236-82-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...