Strontium carbonate ≥97% (contains ~1.0-2.5% barium carbonate)

Supplier: STREM CHEMICALS INC MS

93-3804-1000G 93-3804-250G
100196-636EA 108.98 USD
100196-636 100204-318
Strontium carbonate ≥97% (contains ~1.0-2.5% barium carbonate)
Strontium carbonate
Formula: CO₃Sr
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1497 °C
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR