Iron(II) sulfide ≥99% (Fe basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Ferrous sulphide, Ferrous sulfide

93-2640-100G 93-2640-500G 93-2640-2000G
100196-574EA 175.51 USD
100196-574 100198-346 100203-428
Iron(II) sulfide ≥99% (Fe basis)
Iron (II) sulphide
Formula: FeS
Melting Pt: 1195 °C
Density: 4.694 g/cm³ (20 °C)
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 12,04106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR