Chromium(III) fluoride, anhydrous ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Chromium trifluoride
93-2434-25G
93-2434-1G
93-2434-5G
100208-142EA
992.18
USD
100208-142
100197-762
100196-558
Chromium(III) fluoride, anhydrous ≥98%
Chromium (III) fluoride
Formula:
CrF₃ Melting Pt: > 1000 °C Density: 3.8 g/cm³ (20 °C) |
MDL Number:
MFCD00016045 CAS Number: 7788-97-8 EINECS: 232-137-9 UN: 1756 ADR: 8,II Merck Index: 12,02279 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...