Bis(2-ethylhexyl)phosphate (contains some mono)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

93-1520-500G 93-1520-100G
100199-494EA 230.71 USD
100199-494 100196-514
Bis(2-ethylhexyl)phosphate (contains some mono)
Bis(2-ethylhexyl) phosphate
Formula: C₁₆H₃₅O₄P
Density: 0.974
MDL Number: MFCD00009492
CAS Number: 298-07-7
UN: 1902
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR