Sodium niobate ≥99.9% (Nb basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Sodium niobium oxide

93-1057-5G 93-1057-25G
100197-654EA 65.11 USD
100197-654 100196-448
Sodium niobate ≥99.9% (Nb basis)
Sodium niobate
Formula: NaNbO₃
Melting Pt: 1250 °C
Density: 4,42 g/cm³ (25 °C)
MDL Number: MFCD00014230
CAS Number: 12034-09-2
EINECS: 234-798-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR