Ammonium hydrogen difluoride ≥96% (contains 3-4% NH4F)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0205-500G
93-0205-2000G
100196-412EA
63.69
USD
100196-412
100199-460
Ammonium hydrogen difluoride ≥96% (contains 3-4% NH4F)
Ammonium hydrogen difluoride
Formula:
NH₄HF₂ Melting Pt: 125.6 °C Density: 1.50 Storage Temperature: Ambient |
MDL Number:
MFCD00012018 CAS Number: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II Merck Index: 13,00498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...