Potassium hexachloroplatinate(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dipotassium hexachloroplatinate

78-1960-5G 78-1960-1G
100196-394EA 755.82 USD
100196-394 100196-392
Potassium hexachloroplatinate(IV) ≥99%
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
Melting Pt: 250 °C (dec.)
Density: 3.499
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1,I
Merck Index: 13,07720

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR