Silver ≥99.999%, needles

Supplier: STREM CHEMICALS INC MS

47-0058-10G 47-0058-250G 47-0058-50G
100206-730EA 128.8 USD
100206-730 100196-290 100206-732
Silver ≥99.999%, needles
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR