(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1R,2R)-(-)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese (III) chloride, (R,R)-(-)-N,N'-Bis(3,5-di-tertbutylsalicylidene)-1,2- cyclohexanediaminomanganese(III) chloride, (R,R)-Jacobsen Cat.

25-0300-5G 25-0300-1G
100203-054EA 257.6 USD
100203-054 100196-164
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride

Jacobsen Ligand

Formula: C₃₆H₅₂ClMnN₂O₂
Melting Pt: 324…326 °C
MDL Number: MFCD02101664
CAS Number: 138124-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR