Manganese ≥99%, powder

Supplier: STREM CHEMICALS INC MS
Warning

25-0100-100G 25-0100-500G
100196-160EA 42.46 USD
100196-160 100208-006
Manganese ≥99%, powder
Manganese
Formula: Mn
Boiling Pt: 1900 °C
Melting Pt: 1260 °C
Density: 7.20
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR