Di-p-tolylphosphine ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
15-1820-2G
15-1820-10G
100196-092EA
213.72
USD
100196-092
100196-090
Di-p-tolylphosphine ≥99%
Di-p-tolylphosphine
Phosphine
Formula:
(CH₃C₆H₄)₂PH |
MDL Number:
MFCD01630843 CAS Number: 1017-60-3 UN: 2845 ADR: 4.2,I |
Specification Test Results
Assay | 99% |
Color | Colorless |
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...