Cyclohexyldichlorophosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-0800-1G 15-0800-5G
100196-074EA 108.98 USD
100196-074 100196-076
Cyclohexyldichlorophosphine ≥98%
Cyclohexyldichlorophosphine

Phosphine

Boiling Pt: 100 °C (17 mmHg)
MDL Number: MFCD00014295
CAS Number: 2844-89-5
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR