Dimethoxy(diphenyl)silane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dimethoxydiphenylsilane, Diphenyldimethoxysilane

14-2500-250G 14-2500-50G
100198-120EA 230.71 USD
100198-120 100196-044
Dimethoxy(diphenyl)silane ≥97%
Dimethoxy(diphenyl)silane
Formula: C₁₄H₁₆O₂Si
Boiling Pt: 161 °C (15 mmHg)
Density: 1,0771
Flash Pt: 250 °F
MDL Number: MFCD00025690
CAS Number: 6843-66-9
EINECS: 229-929-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR