Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

56-8600-5G 56-8600-1G
100195-920EA 1535.69 USD
100195-920 100195-918
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct ≥99.99% (Ba basis) (Sr-0.5%), PURATREM
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium tetraglyme adduct

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Melting Pt: 103 °C
MDL Number: MFCD01073799
CAS Number: 136629-60-2
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR