Anti-RNA Polymerase II RPB3 Mouse Monoclonal Antibody [clone: 1Y26]

Supplier: Biolegend
665004
10019-926EA 363.7 USD
10019-926
Anti-RNA Polymerase II RPB3 Mouse Monoclonal Antibody [clone: 1Y26]
Antibodies
Purified anti-RNA Polymerase II RPB3 [1Y26]; Isotype: Mouse IgG1; Reactivity: Yeast (Saccharomyces cerevisiae); Apps: WB, ELISA, ChIP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: RNA Polymerase II RPB3
Clonality: Monoclonal
Clone: 1Y26
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Yeast (Saccharomyces cerevisiae)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR