Anti-RNA Polymerase beta Mouse Monoclonal Antibody [clone: 8RB13]

Supplier: Biolegend
663903
10019-900EA 419.7 USD
10019-900
Anti-RNA Polymerase beta Mouse Monoclonal Antibody [clone: 8RB13]
Antibodies
Anti-E. coli RNA Polymerase β [8RB13]; Isotype: Mouse IgG1; Reactivity: E. coli, Bordetella, Chlamydiae, Myxococcus, Salmonella, Vibrio; Apps: WB, ELISA, IP; Size: 100 μl

Each lot of this antibody is quality control tested by Western blotting. To reconstitute, add 100 μl of ddH2O to the tube containing antibody and mix to dissolve. For Western blotting, the suggested dilution is 1:1000 after reconsitution. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: RNA Polymerase beta
Clonality: Monoclonal
Clone: 8RB13
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: E. coli;Bordetella;Chlamydiae;Myxococcus;Salmonella;Vibrio
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR