Michele Trephine 6.25", Roboz
Supplier: Roboz Surgical
RS-9200
100189-644EA 249.23 USD
100189-644
Michele Trephine 6.25", Roboz
Dissection Sets
Michele Trephine; Graduations 0.25" To 1"; 6.25" Shaft Length; 3/16" Shaft O.D.
These items are designed for animal research only and are not intended for human use.