Dumont #5 Forceps, Dumoxel, Tip Size 0.05×0.01 mm, Biologie Tips, Roboz
Supplier: Roboz Surgical
RS-5060
100189-598EA 65.08 USD
100189-598
Dumont #5 Forceps, Dumoxel, Tip Size 0.05×0.01 mm, Biologie Tips, Roboz
Forceps
Dumont Tweezers; Pattern #5; .10 x .06 mm Tip Size; 110 mm Length; Material: Dumoxel; Biologie Tips