Goldstein Retractor 1" 3×3 4 mm Sharp Prongs, Roboz Surgical
Supplier: Roboz Surgical
RS-6502
100189-164EA 227.08 USD
100189-164
Goldstein Retractor 1" 3×3 4 mm Sharp Prongs, Roboz Surgical
Retractors
Goldstein Retractor; 3 x 3 Prongs; Sharp  Prongs; 4 mm Depth; 19 mm Spread; 1" Length