Micro Vascular Clip, 1.5×5 mm jaws, Roboz Surgical
Supplier: Roboz Surgical
RS-6472
100188-904EA 168.92 USD
100188-904
Micro Vascular Clip, 1.5×5 mm jaws, Roboz Surgical
Animal Supplies Animal Clippers
Micro vascular clip used with forceps.
  • 1 to 2 mm vessel diameter
  • 35 to 40 g occlusion pressure
Caution: These items are designed for animal research only and are not intended for human use.