Micro Vascular Approximator with Moveable Clips 11 mm , Roboz Surgical
Supplier: Roboz Surgical
RS-6490
100188-884EA 535.85 USD
100188-884
Micro Vascular Approximator with Moveable Clips 11 mm , Roboz Surgical
Animal Supplies Animal Clippers
Micro Vascular Approximator With Moveable Clips; 1-2 mm Vessel Diameter; 35-40 Grams Occlusion Pressure; 11 mm Length.
These items are designed for animal research only and are not intended for human use.