Carbide Inverted Cone Burr Size 35, Hand Piece, Roboz Surgical
Supplier: Roboz Surgical
RS-6282C-35
100188-850BX 45.69 USD
100188-850
Carbide Inverted Cone Burr Size 35, Hand Piece, Roboz Surgical
Dissection Sets
Carbide Inverted Cone Burrs; Hand Piece; Size #35, 1 mm Tip; 1.75" Length; 2.33 mm Shaft Diameter; Pack Of 5.
These items are designed for animal research only and are not intended for human use.