Carbide Round Burrs Size 8, Hand Piece, Roboz Surgical
Supplier: Roboz Surgical
RS-6280C-8
100188-846BX 45.69 USD
100188-846
Carbide Round Burrs Size 8, Hand Piece, Roboz Surgical
Dissection Sets
Round Burrs; Hand Piece; Size #8, 2.3 mm Tip; 1.75" Length; 2.33 mm Shaft Diameter; Pack Of 5 For use with the RS-6300 Ideal Micro Drill
These items are designed for animal research only and are not intended for human use.