Micro Trephine 1.8 mm Tungsten Carbide, Roboz Surgical
Supplier: Roboz Surgical
RS-6306
100188-824EA 95.54 USD
100188-824
Micro Trephine 1.8 mm Tungsten Carbide, Roboz Surgical
Dissection Sets
Micro Dissecting Trephine; Tungsten Carbide; 1.8 mm Diameter
The head diameter of these trephines is 1.8 mm. The shaft diameter is 2.33 mm. They work with our Ideal micro drill or other handpieces or drills accepting drill bits with 2.33 mm shaft diameter.
These items are designed for animal research only and are not intended for human use.