Corning® BioCoat™ Pre-Coated PAMPA Plate System, Corning
Supplier: Corning
353015
10018-852CS 886.09 USD
10018-852
Corning® BioCoat™ Pre-Coated PAMPA Plate System, Corning
Microplates Filter Plates
A 96-well, pre-coated filter plate, individually packaged with a 96-well microassay receiver plate.
  • 96-well insert system with a filter plate pre-coated with structured layers of phospholipids
  • Configuration complies with ANSI standards
  • Polystyrene plate with 0.4 µm Polyvinylidene fluoride membrane