Polyvinylfluoride Labels, Brady
Supplier: Brady Worldwide
143579
10018-438EA 1228.53 USD
10018-438
Polyvinylfluoride Labels, Brady
Labels
Durable B-437 Top coated Polyvinyl fluoride material is used most frequently in wire and cable identification applications where self-extinguishing features are required.
  • White or yellow labels feature a matte finish and are thermal transfer printable
  • Material utilizes a permanent acrylic adhesive and has excellent resistance to water, oil, and other common fluids
  • Meets the requirements of MIL-M-87958, Pressure Sensitive Adhesive Wire or Cable Marker and Identification specification
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll