Actinomycin D ≥98% (by HPLC)

Supplier: Biotium

Danger

40038
10018-186EA 191.18 USD
10018-186
Actinomycin D ≥98% (by HPLC)
Actinomycin D

Actinomycin D is a nonfluorescent GC-selective intercalator.


  • Absorption/Emission 240nm & 443nm/none
  • Extinction Coefficient >35,000(240nm) & >25,000(443nm)
  • Orange red solid soluble in DMSO at 50 mg/mL
  • Store at 4°C


Similar to 7-AAD (cat. no. 40038), actinomycin D has also been used for chromosome banding studies.

Formula: C₆₂H₈₆N₁₂O₁₆
MW: 1255.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005033
CAS Number: 50-76-0
EINECS: 200-063-6
UN: 3462
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR