Ultra Durable High Temperature Polyimide Labels, Type B30, UltraTemp®, Brady
Supplier: Brady Worldwide
145296
10018-126EA 1963.78 USD
10018-126
Ultra Durable High Temperature Polyimide Labels, Type B30, UltraTemp®, Brady
Labels
Amber high temperature polyimide labels with an ultra durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component preprocess labeling
  • Resists high temperatures up to 300 °C (572 °F)
  • Fulfills requirements for MIL-STD-202G Method 215K and SAE AS81531 Marking of Electrical Insulating Material when used with R4300 series ribbons
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll