Ultra Aggressive Metalized Polyester Labels, Type B30, MetaLabel™, Brady
Supplier: Brady Worldwide
145294
10018-038EA 390.57 USD
10018-038
Ultra Aggressive Metalized Polyester Labels, Type B30, MetaLabel™, Brady
Labels
Ultra Aggressive Metalized Polyester (B-486) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols, and logos, requiring nameplate-like quality.
  • Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable
  • Designed for high adhesion to textured metals, powder coated surfaces, and low energy plastics
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Best for application to smooth and highly textured surfaces indoors. Cut multiple labels out of a single 4" wide material using BBP®37 Printer.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.