Brady® B30 Series Printer Ribbon R6700 Series
Supplier: Brady Worldwide
133357
10018-014EA 97.58 USD
10018-014
Brady® B30 Series Printer Ribbon R6700 Series
Printer Ribbons
B30 Series "Drop & Click" print ribbons enable 5 second ribbon supply changes.
  • Single color ribbon for printing a single color onto a label
  • R6700 Series resin/wax formulation is a white ribbon that offers good chemical resistance when printed on the appropriate label material from Brady®
  • Recommended for printing on dark-colored heat shrinkable tubing materials, such as black B-342 and B-344 PermaSleeve® wire markers from Brady®
  • Self-contained B30 Series cartridges drop straight into the printer with a "click"
R6700 White is intended for use with black wiremarking sleeves, especially B-344 Fluid Resistant (FR) sleeves).
No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.