Rough Surface Polyester Labels, Type B3, ToughBond®, Brady
Supplier: Brady Worldwide
145292
10017-992EA 315.37 USD
10017-992
Rough Surface Polyester Labels, Type B3, ToughBond®, Brady
Labels
Rough Surface Polyester (B-432) labels are ideal for electronic PCB and component identification, barcode labels, rating plates, and solar panel identification.
  • Made from durable BradyBondz™ glossy white polyester with permanent acrylic adhesive, labels are chemical, oil, and water resistant
  • UL Recognized for outdoor use on glass, thermoset polyester plastic, and polyvinyl fluoride plastic surfaces to support solar panel identification
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Clear polyester material with aggressive permanent adhesive for labeling products and general identification. Best for application to smooth and textured surfaces indoors. Cut multiple labels out of a single 4" wide material using BBP®37 Printer.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; CSA accepted