VACUETTE® Blood Culture Holder, Greiner Bio-One
Supplier: Greiner Bio-One
450181
10017-802CS 326.61 USD
10017-802
VACUETTE® Blood Culture Holder, Greiner Bio-One
Blood Collection Tube Holders
The VACUETTE® Blood Culture Holder is used to fill blood into blood culture bottles and tubes in routine venipuncture procedures.
  • Single-use
  • Nonsterile
  • Plastic construction
  • Compatible with all VACUETTE® Luer adapters
This device is to be used by properly trained healthcare professionals only in accordance with these instructions.
The VACUETTE® Blood Culture Holder is single-use, nonsterile, and made out of plastic. It is compatible with all VACUETTE® Luer Adapters and VACUETTE® Safety/Blood Collection Sets.