SPERM-CHEX® Sperm Count Control, Streck
Supplier: STRECK LABORATORIES
230202
10017-776EA 387.34 USD
10017-776
SPERM-CHEX® Sperm Count Control, Streck
Controls and Calibration Verification Materials
A procedural control with two levels of stabilized sperm cells to validate manual sperm count processes.
  • Comprehensive
  • Two clinically significant levels of control
  • The only sperm count control on the market that contains sperm cells
  • Mimics a patient sample
The control has a 12-month closed-vial stability and a 42-day open-vial stability.