Donor Horse Serum Gamma Irradiated
Supplier: Rocky Mountain Biologicals
DES-BTG-5XM
10015-630EA 96.74 USD
10015-630
Donor Horse Serum Gamma Irradiated
Sera (Serum)
Donor horse serum from RMBIO is derived from US-origin, veterinary-inspected and controlled herds
RMBIO obtains donor horse serum from closed herds that are carefully controlled and inspected by veterinarians. Horses are regularly tested for equine infectious anemia using Coggins tests. Equine serum is a cost-effective alternative to fetal bovine serum (FBS) and can be used as cell culture medium in a variety of settings. 
Equine serum is ideal for use as cell culture medium and in veterinary biologics. This product has been gamma irradiated to meet the needs of our customers.