Cell Cleaning Brushes for ASE 100/150/200/300/350 Cells, Restek
Supplier: Restek
23999
10015-358PK 41.32 USD
10015-358
Cell Cleaning Brushes for ASE 100/150/200/300/350 Cells, Restek
Brushes
Precision cell cleaning brushes produced by Restek are durable and efficient at removing solid samples from cells.
  • Firm nylon bristle brushes for easy cell cleaning and removal of solid samples
  • Range of sizes to fit all extraction cells for ASE 100/150/200/300/350 Cells
Restek cell cleaning brushes are available in a range of sizes for compatibility with various ASE® cells. Nylon brushes are designed to hold up well for quality cell cleaning.