Retaining Ring Plier for ASE® 100/200/300 Systems, Restek
Supplier: Restek
26185
10015-294EA 70.66 USD
10015-294
Retaining Ring Plier for ASE® 100/200/300 Systems, Restek
Pliers
Durable, ergonomically designed pliers are effective at removing retaining rings, both internal and external.
  • Can be used for internal or external retaining rings
  • Works to remove retaining ring in all ASE® cell caps
For use with ASE® 100/200/300 systems.