Reagent alcohol 100%, Surgipath® ISO for histology and pathology

Supplier: Leica Biosystems
Danger

Synonyms: Reagent alcohol

3803686
10015-099CS 162.71 USD
10015-099
Reagent alcohol 100%, Surgipath® ISO for histology and pathology
Alcohol

Ethanols are pure filtered and laboratory tested to ensure a consistent high quality for all formulating, processing and staining procedures in histology and cytology.


  • High-quality filtered Ethanol blend
  • ISO certified
  • Available in non‑breakable, gallon containers


Certifications: ISO certified

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR