Xylene (mixture of isomers), Surgipath®, purified

Supplier: Leica Biosystems
Danger

3803665
10015-093CS 376.02 USD
10015-093
Xylene (mixture of isomers), Surgipath®, purified
Xylene (mixture of isomers)

Purified Xylenes are highly refined, pure filtered and laboratory tested to ensure consistent high quality for all histology and cytology procedures.


  • High-quality filtered xylene
  • ISO certified
  • Available in non‑breakable, solvent‑resistant containers


Aromatic hydrocarbon used for Histological tissue processing and Hstological/Cytoloical staining procedures.


Certifications: ISO certified

Formula: C₆H₄(CH₃)₂
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR