Eosin Y (yellowish) alcoholic solution, Surgipath® ready-to-use, used in nuclear staining

Supplier: Leica Biosystems

3801600
10015-072EA 287.47 USD
10015-072
Eosin Y (yellowish) alcoholic solution, Surgipath® ready-to-use, used in nuclear staining
Eosin Y (yellowish)

Eosin Secondary-Counter Stains


Eosin Y is an alcohol-based, ready-to-use secondary stain used primarily in histology for routine hematoxylin and eosin staining.


It may also be used in cytology for fast cytoplasmic staining of rush specimens also using the H&E technique.

Formula: C₂₀H₆Br₄Na₂O₅
MDL Number: MFCD00005040
CAS Number: 17372-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR