Hematoxylin solution Gill II, Surgipath® used in nuclear staining mercury-free

Supplier: Leica Biosystems

3801520
10015-068GL 172.03 USD
10015-068 75810-402
Hematoxylin solution Gill II, Surgipath® used in nuclear staining mercury-free
Hematoxylin staining solutions

The Gill formula hematoxylins are used in the progressive method of staining.


Three Gill formulations are available:
The Gill I formula is generally recommended for cytology.
The stronger Gill III is usually preferred for use in histology.
GIll II is a medium strength hematoxylin.

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR