Micro Slides, Leica
Supplier: Leica Biosystems
3800240
10014-958CS 498.65 USD
10014-958
Micro Slides, Leica
Slides Microscope Slides
Micro Slides use high-quality sheet glass in a carefully controlled environment, implementing strict specifications and quality control procedures.
  • Precleaned
  • Packed in environmentally friendly cardboard boxes
  • Frosted ends allow easy marking and identification
Each slide is precleaned, inspected four separate times, and packed in environmentally friendly cardboard boxes. Frosted end facilitates marking and identification.