Orion™ AQUAfast AQ3010 Turbidity Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
AQ3010
10014-932KT 1384.43 USD
10014-932
Orion™ AQUAfast AQ3010 Turbidity Meter, Thermo Scientific
Turbidity Meters
Generate an accurate turbidity reading quickly with the Orion™ AQUAfast AQ3010 Turbidity Meter.
  • Easy to read results from the large, bright LCD display in a variety of work environments
  • Autoranging from 0 to 1000 NTUs
  • Meter uses Nephelometric principle with 90° detection
  • Meets ISO 7027 and offers consistent light source for greater accuracy
  • No mixing or diluting with EPA-approved polymer bead primary standards for faster calibration
Perfect for use in the plant, field, or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life, averaging approximately 3000 tests.
Certifications: CE, TUV 3-in-1 for CSA-US, FCC part 15.
Ordering information: Replacement products include: Parts Set AC301S, Silicone Oil and Cloth AC3SIL, and Vial AC3V25. Other accessories include Cleaning Brush AC3CBR and Formazin Turbidity Standard (4000 NTU, 475mL) AC45FZ. All accessories and replacement parts are sold separately. Subject to all warranty requirements and exclusions set out in the Terms and Conditions of Sale, this product is warranted from the date shipped and for ninety (90) days thereafter.