Shaker Flask Clamps, Triforest Enterprises
Supplier: TRIFOREST ENTERPRISES INC
SFC0125
10014-814EA 62.84 USD
10014-814 10014-816 10014-818 10014-854 10014-856
Shaker Flask Clamps, Triforest Enterprises
Clamps General Purpose Clamps
Polypropylene shaker clamps are carefully engineered to provide both clipping elasticity and motion resistance in one product
Featuring a non-scratch design, these shaker clamps come in standard blue color accompanied by stainless steel screws and washers. Clamps fit all industry standard shaking incubators and are autoclavable for reuse.