AMPIGENE® Taq DNA Polymerase, Enzo Life Sciences
Supplier: Enzo Life Sciences
PRT100-0500
10014-680EA 163.5 USD
10014-680
AMPIGENE® Taq DNA Polymerase, Enzo Life Sciences
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Optimized standard PCR for enhanced speed, yield, and specificity.
For standard PCR applications.
AMPIGENE® Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. It can be used for routine PCR applications including genotyping, screening, and library construction.
Ordering information: Contains AMPIGENE® Taq DNA polymerase (5u/µl)and 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).