Sodium chloride 0.9% Physiological saline endotoxin-free, sterile

Supplier: Enzo Life Sciences

505-009-LD15
10014-564EA 58.86 USD
10014-564
Sodium chloride 0.9% Physiological saline endotoxin-free, sterile
Sodium chloride

Sterile and endotoxin-free physiological saline for use with TLRgrade™ or endotoxin-free grade reagents.


Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal.

Formula: NaCl
Storage Temperature: Refrigerator
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Endotoxin content <0.0002 EU/µg (pyrotell gel-clot endotoxin test)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR