POLYVIEW® (Mouse on Mouse-HRP) IHC Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
950-114-0100
10014-544EA
924.32
USD
10014-544
POLYVIEW® (Mouse on Mouse-HRP) IHC Kit, Enzo Life Sciences
Assays
Immunological Assays
Highest sensitivity, lowest background nanopolymer detection reagent for use with HIGHDEF® chromogens.
Ordering information: Contains IHC tissue primer, blocking reagent, anti-mouse HRP polymer, DAB buffer, DAB chromogen, empty mixing bottle for DAB.
Caution: RUO - Research Use Only