AMPIGENE® DNA Ladder 250 to 10000 bp, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN103-0100
10014-498EA 98.1 USD
10014-498
AMPIGENE® DNA Ladder 250 to 10000 bp, Enzo Life Sciences
Electrophoresis Reagents Molecular Weight Markers
Ready to use DNA standards for quantifying PCR yield
For size determination and DNA quantification using agarose electrophoresis
AMPIGENE® DNA Ladder 250-10,000 bp contains DNA fragments ranging from 250bp to 10,000bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 1,000bp and 3,000bp to serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5µl load, for determining the approximate mass of DNA in samples with comparable intensity and size.
Ordering information: Contains Ready-to-use AMPIGENE® DNA Ladder 250-10,000 bp and Sample Loading Buffer.